Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194778.2 | NP_001181707.1 | 245 | 77 | ||
GenBank mRNA | JU324736.1 | - | 237 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |