Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2CD4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194820.2 | NP_001181749.1 | 1233 | 89 | ||
GenBank mRNA | JV565065.1 | - | 1233 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |