Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR4Q3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194892.1 | NP_001181821.1 | 846 | 101 | ||
XM_015143076.2 | XP_014998562.1 | 1005 | 101 | |||
XM_015143079.2 | XP_014998565.1 | 1019 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |