Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190845.1 | NP_001177774.1 | 129 | 90 | ||
GenBank mRNA | CB554885.1 | - | 129 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |