Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190876.1 | NP_001177805.1 | 241 | 62 | ||
XM_015132068.2 | XP_014987554.1 | 618 | 62 | |||
XM_028843441.1 | XP_028699274.1 | 673 | 62 | |||
XM_028843442.1 | XP_028699275.1 | 520 | 62 | |||
GenBank mRNA | JU335721.1 | - | 296 | 62 | ||
JU476609.1 | - | 321 | 62 | |||
JV589438.1 | - | 254 | 62 | |||
JV691972.1 | - | 255 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |