Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GXYLT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193691.3 | NP_001180620.1 | 1146 | 79 | ||
XM_028843068.1 | XP_028698901.1 | 1180 | 79 | |||
GenBank mRNA | JU492654.1 | - | 208 | 79 | ||
JV070198.1 | - | 952 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |