Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199128.1 | NP_001186057.1 | 820 | 76 | ||
GenBank mRNA | JV584466.1 | - | 394 | 76 |
Gene Symbol: PHKG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU476003.1 | - | 1810 | 76 | ||
JV653731.1 | - | 1857 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |