Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204311.2 | NP_001191240.2 | 252 | 65 | ||
XM_028840325.1 | XP_028696158.1 | 273 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |