Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DECR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204462.2 | NP_001191391.1 | 668 | 74 | ||
XM_015125157.2 | XP_014980643.1 | 685 | 74 | |||
XM_015125158.2 | XP_014980644.1 | 687 | 74 | |||
XM_028840375.1 | XP_028696208.1 | 684 | 74 | |||
GenBank mRNA | JV057526.1 | - | 666 | 74 | ||
JV635589.1 | - | 663 | 74 | |||
JV668438.1 | - | 640 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |