Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204518.1 | NP_001191447.1 | 403 | 82 | ||
XM_015119231.2 | XP_014974717.1 | 568 | 82 | |||
XM_028835441.1 | XP_028691274.1 | 444 | 82 | |||
GenBank mRNA | CO581875.1 | - | 407 | 82 | ||
JV600817.1 | - | 572 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |