Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194593.2 | NP_001181522.1 | 303 | 115 | ||
GenBank mRNA | JU333891.1 | - | 367 | 115 | ||
JU475076.1 | - | 311 | 115 | |||
JV044876.1 | - | 320 | 115 | |||
JV669285.1 | - | 378 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |