Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194593.2 | NP_001181522.1 | 776 | 109 | ||
GenBank mRNA | JU333891.1 | - | 840 | 109 | ||
JU475076.1 | - | 784 | 109 | |||
JV044876.1 | - | 793 | 109 | |||
JV669285.1 | - | 851 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |