Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194822.2 | NP_001181751.2 | 359 | 95 | ||
XM_001104117.4 | XP_001104117.2 | 533 | 95 | |||
XM_015122911.2 | XP_014978397.2 | 650 | 95 | |||
GenBank mRNA | JU472854.1 | - | 510 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |