Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194893.2 | NP_001181822.1 | 423 | 105 | ||
GenBank mRNA | JU337017.1 | - | 201 | 105 | ||
JU472497.1 | - | 226 | 105 | |||
JV525112.1 | - | 99 | 105 | |||
JV667681.1 | - | 232 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |