Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260699.1 | NP_001247628.1 | 565 | 84 | ||
XM_015143632.2 | XP_014999118.1 | 664 | 84 | |||
GenBank mRNA | JU319907.1 | - | 522 | 84 | ||
JU476838.1 | - | 565 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |