Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243275.3 | NP_001230204.2 | 297 | 67 | ||
XM_028827327.1 | XP_028683160.1 | 306 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |