Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPLP0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195428.1 | NP_001182357.1 | 832 | 69 | ||
XM_015152909.2 | XP_015008395.1 | 897 | 69 | |||
GenBank mRNA | AF162483.1 | - | 133 | 69 | ||
CN642741.1 | - | 617 | 69 | |||
CN643487.1 | - | 832 | 69 | |||
JV047873.1 | - | 863 | 69 | |||
JV625067.1 | - | 877 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Endogenous Controls Recommended Assay Macaca mulatta |