Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257304.2 | NP_001244233.2 | 46 | 102 | ||
GenBank mRNA | HQ423396.1 | - | 35 | 102 | ||
JQ037994.2 | - | 46 | 102 | |||
JQ037995.1 | - | 46 | 102 | |||
JQ037996.2 | - | 46 | 102 | |||
JU333388.1 | - | 80 | 102 | |||
JV489157.1 | - | 52 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |