Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257418.1 | NP_001244347.1 | 135 | 70 | ||
GenBank mRNA | JU321633.1 | - | 135 | 70 | ||
JU470880.1 | - | 112 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |