Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257418.1 | NP_001244347.1 | 282 | 97 | ||
XM_015140045.2 | XP_014995531.1 | 704 | 97 | |||
XM_015140047.2 | XP_014995533.1 | 388 | 97 | |||
XM_015140048.2 | XP_014995534.1 | 387 | 97 | |||
XM_015140050.2 | XP_014995536.1 | 705 | 97 | |||
XM_028849230.1 | XP_028705063.1 | 2063 | 97 | |||
GenBank mRNA | JU321633.1 | - | 282 | 97 | ||
JU470880.1 | - | 259 | 97 |