Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257888.2 | NP_001244817.1 | 754 | 92 | ||
XM_015149824.2 | XP_015005310.1 | 858 | 92 | |||
GenBank mRNA | JU336021.1 | - | 781 | 92 | ||
JU336022.1 | - | 754 | 92 | |||
JU476885.1 | - | 738 | 92 | |||
JU476886.1 | - | 711 | 92 | |||
JV047994.1 | - | 804 | 92 | |||
JV047995.1 | - | 777 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |