Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260699.1 | NP_001247628.1 | 509 | 127 | ||
XM_015143632.2 | XP_014999118.1 | 608 | 127 | |||
GenBank mRNA | JU319907.1 | - | 466 | 127 | ||
JU476838.1 | - | 509 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |