Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYKPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260930.2 | NP_001247859.1 | 1203 | 85 | ||
GenBank mRNA | JU322930.1 | - | 1203 | 85 | ||
JU478189.1 | - | 1277 | 85 |