Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261331.1 | NP_001248260.1 | 426 | 69 | ||
GenBank mRNA | JU336099.1 | - | 426 | 69 |
Gene Symbol: TAPBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU492265.1 | - | 415 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |