Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261480.1 | NP_001248409.1 | 1018 | 80 | ||
GenBank mRNA | JU326985.1 | - | 364 | 80 | ||
JU477353.1 | - | 1018 | 80 | |||
JV594909.1 | - | 667 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |