Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261503.1 | NP_001248432.1 | 680 | 71 | ||
XM_015150747.2 | XP_015006233.1 | 71 | ||||
GenBank mRNA | JU335998.1 | - | 680 | 71 | ||
JV703582.1 | - | 1443 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |