Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBNDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261592.1 | NP_001248521.1 | 351 | 90 | ||
GenBank mRNA | JU332295.1 | - | 351 | 90 | ||
JV557468.1 | - | 323 | 90 |