Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBEY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265798.1 | NP_001252727.1 | 510 | 66 | ||
XM_015132652.2 | XP_014988138.1 | 856 | 66 | |||
XM_015132654.2 | XP_014988140.1 | 396 | 66 | |||
GenBank mRNA | JU323926.1 | - | 492 | 66 | ||
JV636225.1 | - | 510 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |