Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265960.1 | NP_001252889.1 | 166 | 79 | ||
GenBank mRNA | JU336223.1 | - | 152 | 79 | ||
JU477060.1 | - | 165 | 79 | |||
JV048150.1 | - | 166 | 79 | |||
JV602090.1 | - | 278 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |