Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASTOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266104.1 | NP_001253033.1 | 692 | 113 | ||
GenBank mRNA | JU328414.1 | - | 639 | 113 | ||
JU477044.1 | - | 692 | 113 | |||
JV480559.1 | - | 647 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |