Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASTOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266104.1 | NP_001253033.1 | 971 | 85 | ||
GenBank mRNA | JU328414.1 | - | 918 | 85 | ||
JU477044.1 | - | 971 | 85 | |||
JV480559.1 | - | 926 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |