Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266415.1 | NP_001253344.1 | 104 | 74 | ||
GenBank mRNA | JU323492.1 | - | 100 | 74 | ||
JU473824.1 | - | 104 | 74 | |||
JV043222.1 | - | 104 | 74 | |||
JV529264.1 | - | 81 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |