Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266473.1 | NP_001253402.1 | 1618 | 68 | ||
XM_015144264.2 | XP_014999750.1 | 2448 | 68 | |||
XM_015144265.2 | XP_014999751.1 | 3126 | 68 | |||
GenBank mRNA | JU319924.1 | - | 2038 | 68 | ||
JV043836.1 | - | 1618 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |