Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266511.1 | NP_001253440.1 | 427 | 111 | ||
GenBank mRNA | JU335172.1 | - | 433 | 111 | ||
JU472074.1 | - | 422 | 111 | |||
JV047473.1 | - | 427 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |