Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266547.1 | NP_001253476.1 | 557 | 97 | ||
GenBank mRNA | JU335565.1 | - | 589 | 97 | ||
JV045033.1 | - | 557 | 97 | |||
JV656521.1 | - | 587 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |