Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301712.1 | NP_001288641.1 | 422 | 76 | ||
GenBank mRNA | JU333004.1 | - | 434 | 76 | ||
JU472798.1 | - | 412 | 76 | |||
JV046232.1 | - | 422 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |