Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266612.1 | NP_001253541.1 | 447 | 112 | ||
XM_015150399.2 | XP_015005885.1 | 430 | 112 | |||
XM_015150400.2 | XP_015005886.1 | 640 | 112 | |||
XM_028827305.1 | XP_028683138.1 | 423 | 112 | |||
XM_028827306.1 | XP_028683139.1 | 423 | 112 | |||
XM_028827307.1 | XP_028683140.1 | 423 | 112 | |||
GenBank mRNA | JU333220.1 | - | 546 | 112 | ||
JU474539.1 | - | 550 | 112 | |||
JV046339.1 | - | 447 | 112 | |||
JV573333.1 | - | 537 | 112 | |||
JV639477.1 | - | 569 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |