Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266626.2 | NP_001253555.1 | 730 | 72 | ||
XM_015134612.2 | XP_014990098.1 | 1131 | 72 | |||
XM_015134614.2 | XP_014990100.1 | 1442 | 72 | |||
XM_015134615.2 | XP_014990101.1 | 1051 | 72 | |||
XM_015134616.2 | XP_014990102.1 | 988 | 72 | |||
XM_015134617.2 | XP_014990103.1 | 978 | 72 | |||
XM_015134618.2 | XP_014990104.1 | 1345 | 72 | |||
XM_028844850.1 | XP_028700683.1 | 1021 | 72 | |||
XM_028844851.1 | XP_028700684.1 | 1128 | 72 | |||
XM_028844852.1 | XP_028700685.1 | 1048 | 72 | |||
GenBank mRNA | JU327681.1 | - | 655 | 72 | ||
JU471738.1 | - | 732 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |