Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREBZF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266648.1 | NP_001253577.1 | 822 | 138 | ||
XM_015115294.2 | XP_014970780.1 | 854 | 138 | |||
XM_028832600.1 | XP_028688433.1 | 854 | 138 | |||
GenBank mRNA | JU335725.1 | - | 840 | 138 | ||
JU335726.1 | - | 840 | 138 | |||
JU476610.1 | - | 838 | 138 | |||
JU476611.1 | - | 838 | 138 | |||
JV047803.1 | - | 822 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |