Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261294.1 | NP_001248223.1 | 217 | 104 | ||
XM_028846659.1 | XP_028702492.1 | 396 | 104 | |||
XM_028846660.1 | XP_028702493.1 | 485 | 104 | |||
XM_028846661.1 | XP_028702494.1 | 406 | 104 | |||
XM_028846662.1 | XP_028702495.1 | 265 | 104 | |||
GenBank mRNA | JU323501.1 | - | 189 | 104 | ||
JU336563.1 | - | 189 | 104 | |||
JU471488.1 | - | 183 | 104 | |||
JV043650.1 | - | 217 | 104 | |||
JV043651.1 | - | 217 | 104 | |||
JV489265.1 | - | 264 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |