Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABARAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260686.1 | NP_001247615.1 | 280 | 129 | ||
GenBank mRNA | JU319881.1 | - | 161 | 129 | ||
JU474998.1 | - | 257 | 129 | |||
JV045161.1 | - | 280 | 129 | |||
JV528078.1 | - | 331 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |