Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266471.1 | NP_001253400.1 | 619 | 120 | ||
XM_028827241.1 | XP_028683074.1 | 570 | 120 | |||
GenBank mRNA | JV600297.1 | - | 619 | 120 | ||
JV655644.1 | - | 507 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |