Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM210A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266721.1 | NP_001253650.1 | 414 | 109 | ||
XM_015121547.2 | XP_014977033.1 | 571 | 109 | |||
XM_028837720.1 | XP_028693553.1 | 443 | 109 | |||
XM_028837721.1 | XP_028693554.1 | 697 | 109 | |||
XM_028837722.1 | XP_028693555.1 | 390 | 109 | |||
GenBank mRNA | JU332222.1 | - | 363 | 109 | ||
JV043598.1 | - | 414 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |