Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC721200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136026.2 | XP_014991512.2 | 2143 | 94 |
Gene Symbol: LOC723314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028847707.1 | XP_028703540.1 | 2085 | 94 | ||
GenBank mRNA | JU329550.1 | - | 1996 | 94 | ||
JU471116.1 | - | 2128 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta Inventoried |