Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_132147.3 | 252 | 101 | |||
GenBank mRNA | AY026237.1 | - | 614 | 101 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |