Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053805.1 | NP_446257.1 | 1 - 2 | 162 | 79 | |
GenBank mRNA | BC079210.1 | - | 1 - 2 | 162 | 7131806 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |