Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053338.2 | NP_445790.2 | 2 - 3 | 540 | 71 | |
XM_006255055.2 | XP_006255117.1 | 2 - 3 | 622 | 71 | ||
GenBank mRNA | CK602305.1 | - | 2 - 3 | 512 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |