Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005881.3 | NP_001005881.2 | 9 - 10 | 1575 | 62 | |
XM_006242628.2 | XP_006242690.1 | 9 - 10 | 1960 | 62 | ||
XM_006242629.2 | XP_006242691.1 | 7 - 8 | 1097 | 62 | ||
XM_008765930.1 | XP_008764152.1 | 9 - 10 | 1885 | 62 | ||
XM_008765931.1 | XP_008764153.1 | 8 - 9 | 1771 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |