Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fev | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144753.2 | NP_653354.2 | 2 - 3 | 314 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |