Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpm6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178105.2 | NP_835206.1 | 3 - 4 | 406 | 67 | |
XM_006253093.2 | XP_006253155.1 | 4 - 5 | 1242 | 67 | ||
GenBank mRNA | AB089242.1 | - | 3 - 4 | 385 | 67 | |
BC088862.1 | - | 3 - 4 | 406 | 7313006 | 67 | |
FQ211804.1 | - | 4 - 5 | 861 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |